Boc-Arg(NO₂)-OH
Supplier: Matrix Scientific
Synonyms:
Nα-Boc-Nomega-nitro-L-arginine
041605-5G
041605-25G
101852-446EA
40
USD
101852-446
101852-448
Boc-Arg(NO₂)-OH
Boc-Arg(NO₂)-OH
CAS: 2188-18-3
MDL Number: MFCD00065556
Molecular Formula: C11H21N5O6
Molecular Weight: 319,32
Hazard: Irritant
Formula:
C₁₁H₂₁N₅O₆ MW: 319.318 g/mol |
MDL Number:
MFCD00065556 CAS Number: 2188-18-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...