Boc-Lys(ME)2-OH
Supplier: Matrix Scientific
Synonyms:
Boc-Lys(Me)₂-OH
041603-1G
041603-250MG
101852-440EA
706.15
USD
101852-440
101852-438
Boc-Lys(ME)2-OH
Boc-Lys(ME)2-OH
CAS: 65671-53-6
MDL Number: MFCD00076965
Molecular Formula: C13H26N2O4
Molecular Weight: 274,36
Hazard: Irritant
Formula:
C₁₃H₂₆N₂O₄ MW: 274.361 g/mol |
MDL Number:
MFCD00076965 CAS Number: 65671-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...