(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclopentylacetic acid

Supplier: Matrix Scientific

Synonyms: N-Fmoc-2-cyclopentyl-L-glycineFmoc-Cpg-OH

041557-1G
101852-260EA 175.95 USD
101852-260
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclopentylacetic acid
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-cyclopentylacetic acid

Hazard: Irritant

Formula: C₂₂H₂₃NO₄
MDL Number: MFCD01311751
CAS Number: 220497-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR