Fmoc-L-cyclopropylalanine

Supplier: Matrix Scientific

Synonyms: Fmoc-β-cyclopropyl-L-AlanineFmoc-β-cyclopropyl-Ala-OH

041553-1G
101852-244EA 90.85 USD
101852-244
Fmoc-L-cyclopropylalanine
Fmoc-L-cyclopropylalanine

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MW: 351.402 g/mol
MDL Number: MFCD00798640
CAS Number: 214750-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR