Fmoc-β-chloro-L-alanine

Supplier: Matrix Scientific

041547-1G 041547-5G
101852-216EA 205.85 USD
101852-216 101852-218
Fmoc-β-chloro-L-alanine
Fmoc-beta-chloro-L-alanine

Hazard: Irritant

Formula: C₁₈H₁₆ClNO₄
MDL Number: MFCD00270534
CAS Number: 212651-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR