(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
N-Boc-2-cyclohexyl-L-glycine
, BOC-L-α-cyclohexylglycine
, N-Boc-L-2-cyclohexylglycine
, Boc-Chg-OH
, Boc-cyclohexyl-Gly-OH
041477-5G
041477-1G
101851-936EA
63.08
USD
101851-936
101851-934
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid ≥95%
(S)-2-((tert-Butoxycarbonyl)amino)-2-cyclohexylacetic acid
Hazard: Irritant
Formula:
C₁₃H₂₃NO₄ MW: 257.3 g/mol |
MDL Number:
MFCD00133638 CAS Number: 109183-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...