trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid

Supplier: Matrix Scientific

Synonyms: trans-4-(Boc-amino)cyclohexanecarboxylic Acid

041368-5G
101851-506EA 50.6 USD
101851-506
trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic Acid
trans-4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid

CAS: 53292-89-0
MDL Number: MFCD01862293
Molecular Formula: C12H21NO4
Molecular Weight: 243,31
Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MW: 243.30 g/mol
MDL Number: MFCD01862293
CAS Number: 53292-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR