(1S,2R)-Fmoc-2-amino-1-cyclopentanecarboxylic acid

Supplier: Matrix Scientific

041324-50MG
101851-342EA 818.8 USD
101851-342
(1S,2R)-Fmoc-2-amino-1-cyclopentanecarboxylic acid
(1S,2R)-Fmoc-2-amino-1-cyclopentanecarboxylic acid

Hazard: Irritant

Formula: C₂₁H₂₁NO₄
MDL Number: MFCD09750529
CAS Number: 352707-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR