(1R,2S)-Fmoc-2-aminocyclohex-3-enecarboxylic acid

Supplier: Matrix Scientific

Synonyms: cis-2-(9-Fluorenylmethoxycarbonylamino)cyclohex-3-enecarboxylic acid

041315-50MG
101851-314EA 615.25 USD
101851-314
(1R,2S)-Fmoc-2-aminocyclohex-3-enecarboxylic acid
(1R,2S)-Fmoc-2-aminocyclohex-3-enecarboxylic acid

Hazard: Irritant

Formula: C₂₂H₂₁NO₄
MW: 363.42 g/mol
MDL Number: MFCD09750521
CAS Number: 233600-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR