(1R,2S)-Boc-2-aminocyclohex-3-enecarboxylic acid

Supplier: Matrix Scientific

041310-50MG
101851-302EA 615.25 USD
101851-302
(1R,2S)-Boc-2-aminocyclohex-3-enecarboxylic acid
(1R,2S)-Boc-2-aminocyclohex-3-enecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₉NO₄
MDL Number: MFCD09750520
CAS Number: 870288-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR