cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid

Supplier: Matrix Scientific

Synonyms: cis-2-((tert-Butoxycarbonyl)amino)cyclopentanecarboxylic acid(1R,2S)-2-(Boc-amino)cyclopentanecarboxylic Acid

041308-50MG
101851-298EA 840 USD
101851-298
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid
cis-2-(tert-Butoxycarbonylamino)-1-cyclopentanecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₄
MW: 229.276 g/mol
MDL Number: MFCD00800539
CAS Number: 136315-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR