Fmoc-HomoArg(Pbf)-OH

Supplier: Matrix Scientific

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(3-((2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl)sulfonyl)guanidino)hexanoic acid

041281-1G 041281-5G
101851-184EA 131.1 USD
101851-184 101851-182
Fmoc-HomoArg(Pbf)-OH
Fmoc-HomoArg(Pbf)-OH

Hazard: Irritant

Formula: C₃₅H₄₂N₄O₇S
MW: 662.81 g/mol
MDL Number: MFCD03093571
CAS Number: 1159680-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR