3-(Difluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

041236-1G
101851-004EA 60 USD
101851-004
3-(Difluoromethyl)pyridine ≥97%
3-(Difluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₅F₂N
MW: 129.11 g/mol
MDL Number: MFCD11226584
CAS Number: 76541-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR