2-(4-Benzylpiperazino-1-yl)-6-fluorophenol

Supplier: Matrix Scientific

041172-250MG
101850-940EA 345 USD
101850-940
2-(4-Benzylpiperazino-1-yl)-6-fluorophenol
2-(4-Benzylpiperazino-1-yl)-6-fluorophenol

Hazard: Irritant

Formula: C₁₇H₁₉FN₂O
MDL Number: MFCD11849914
CAS Number: 1171918-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR