2-(4-Boc-piperazino-1-yl)-4-bromophenol

Supplier: Matrix Scientific

041168-250MG
101850-932EA 345 USD
101850-932
2-(4-Boc-piperazino-1-yl)-4-bromophenol
2-(4-Boc-piperazino-1-yl)-4-bromophenol

Hazard: Irritant

Formula: C₁₅H₂₁BrN₂O₃
MDL Number: MFCD11849911
CAS Number: 1171917-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR