2-(4-Boc-piperazino-1-yl)-5-bromophenol

Supplier: Matrix Scientific

041167-250MG
101850-930EA 345 USD
101850-930
2-(4-Boc-piperazino-1-yl)-5-bromophenol
2-(4-Boc-piperazino-1-yl)-5-bromophenol

Hazard: Irritant

Formula: C₁₅H₂₁BrN₂O₃
MDL Number: MFCD11849910
CAS Number: 1171917-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR