4-(4-Benzyl-1-piperazino)-2-fluorophenol

Supplier: Matrix Scientific

041160-250MG
101850-922EA 356.5 USD
101850-922
4-(4-Benzyl-1-piperazino)-2-fluorophenol
4-(4-Benzyl-1-piperazino)-2-fluorophenol

Hazard: Irritant

Formula: C₁₇H₁₉FN₂O
MDL Number: MFCD11974215
CAS Number: 1171917-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR