2-(4-Boc-piperazino-1-yl)-4-bromobenzaldehyde

Supplier: Matrix Scientific

041143-500MG
101850-890EA 320.85 USD
101850-890
2-(4-Boc-piperazino-1-yl)-4-bromobenzaldehyde
2-(4-Boc-piperazino-1-yl)-4-bromobenzaldehyde

Hazard: Irritant

Formula: C₁₆H₂₁BrN₂O₃
MDL Number: MFCD11849903
CAS Number: 1171917-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR