7-Bromo-1-phenylsulfonyl-3-chlorosulfonylindol

Supplier: Matrix Scientific

041137-250MG
101850-878EA 441.6 USD
101850-878
7-Bromo-1-phenylsulfonyl-3-chlorosulfonylindol
7-Bromo-1-phenylsulfonyl-3-chlorosulfonylindol

Hazard: Irritant

Formula: C₁₄H₉BrClNO₄S₂
MDL Number: MFCD11849924
CAS Number: 1171917-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR