((4-Methoxyphenyl){2-oxo-2-[(2-phenylethyl)amino]ethyl}amino)acetic acid

Supplier: Matrix Scientific

039089-500MG
101846-800EA 144.9 USD
101846-800
((4-Methoxyphenyl){2-oxo-2-[(2-phenylethyl)amino]ethyl}amino)acetic acid
((4-Methoxyphenyl){2-oxo-2-[(2-phenylethyl)amino]ethyl}amino)acetic acid

Hazard: Irritant

Formula: C₁₉H₂₂N₂O₄
MDL Number: MFCD12027593
CAS Number: 1142204-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR