[(2-{[2-(3-Chlorophenyl)ethyl]amino}-2-oxoethyl)-(4-methoxyphenyl)amino]acetic acid

Supplier: Matrix Scientific

039088-500MG
101846-798EA 144.9 USD
101846-798
[(2-{[2-(3-Chlorophenyl)ethyl]amino}-2-oxoethyl)-(4-methoxyphenyl)amino]acetic acid
[(2-{[2-(3-Chlorophenyl)ethyl]amino}-2-oxoethyl)-(4-methoxyphenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₉H₂₁ClN₂O₄
MDL Number: MFCD12027592
CAS Number: 1142204-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR