[{2-[(2-Cyclohex-1-en-1-ylethyl)amino]-2-oxoethyl}(4-methoxyphenyl)amino]acetic acid

Supplier: Matrix Scientific

039080-500MG
101846-782EA 144.9 USD
101846-782
[{2-[(2-Cyclohex-1-en-1-ylethyl)amino]-2-oxoethyl}(4-methoxyphenyl)amino]acetic acid
[{2-[(2-Cyclohex-1-en-1-ylethyl)amino]-2-oxoethyl}(4-methoxyphenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₉H₂₆N₂O₄
MDL Number: MFCD12027584
CAS Number: 1142216-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR