((4-Methoxyphenyl){2-oxo-2-[(pyridin-3-ylmethyl)amino]ethyl}amino)acetic acid

Supplier: Matrix Scientific

039079-500MG
101846-780EA 144.9 USD
101846-780
((4-Methoxyphenyl){2-oxo-2-[(pyridin-3-ylmethyl)amino]ethyl}amino)acetic acid
((4-Methoxyphenyl){2-oxo-2-[(pyridin-3-ylmethyl)amino]ethyl}amino)acetic acid

Hazard: Irritant

Formula: C₁₇H₁₉N₃O₄
MDL Number: MFCD12027583
CAS Number: 1142215-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR