[(2-{[2-(3-Fluorophenyl)ethyl]amino}-2-oxoethyl)(4-methoxyphenyl)amino]acetic acid

Supplier: Matrix Scientific

039078-500MG
101846-778EA 144.9 USD
101846-778
[(2-{[2-(3-Fluorophenyl)ethyl]amino}-2-oxoethyl)(4-methoxyphenyl)amino]acetic acid
[(2-{[2-(3-Fluorophenyl)ethyl]amino}-2-oxoethyl)(4-methoxyphenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₉H₂₁FN₂O₄
MDL Number: MFCD12027582
CAS Number: 1142215-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR