[{2-[(4-Fluorobenzyl)amino]-2-oxoethyl}(4-methoxyphenyl)amino]acetic acid

Supplier: Matrix Scientific

039066-500MG
101846-754EA 144.9 USD
101846-754
[{2-[(4-Fluorobenzyl)amino]-2-oxoethyl}(4-methoxyphenyl)amino]acetic acid
[{2-[(4-Fluorobenzyl)amino]-2-oxoethyl}(4-methoxyphenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₈H₁₉FN₂O₄
MDL Number: MFCD12027570
CAS Number: 1142215-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR