[{2-[(1,3-Benzodioxol-5-ylmethyl)amino]-2-oxoethyl}(4-methoxyphenyl)amino]acetic acid

Supplier: Matrix Scientific

039065-500MG
101846-752EA 144.9 USD
101846-752
[{2-[(1,3-Benzodioxol-5-ylmethyl)amino]-2-oxoethyl}(4-methoxyphenyl)amino]acetic acid
[{2-[(1,3-Benzodioxol-5-ylmethyl)amino]-2-oxoethyl}(4-methoxyphenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₉H₂₀N₂O₆
MDL Number: MFCD12027569
CAS Number: 1142215-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR