Ethyl-2-amino-5-phenyl-1,3-thiazole-4-carboxylate hydrochloride

Supplier: Matrix Scientific

039061-500MG
101846-744EA 345 USD
101846-744
Ethyl-2-amino-5-phenyl-1,3-thiazole-4-carboxylate hydrochloride
Ethyl-2-amino-5-phenyl-1,3-thiazole-4-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂O₂S
MDL Number: MFCD28013197
CAS Number: 77505-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR