2,2-Dimethyl-3-[(4-methyl-1,4-diazepan-1-yl)carbonyl]cyclopropanecarboxylic acid

Supplier: Matrix Scientific

039058-500MG
101846-738EA 217.35 USD
101846-738
2,2-Dimethyl-3-[(4-methyl-1,4-diazepan-1-yl)carbonyl]cyclopropanecarboxylic acid
2,2-Dimethyl-3-[(4-methyl-1,4-diazepan-1-yl)carbonyl]cyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₂₂N₂O₃
MDL Number: MFCD12027563
CAS Number: 1142215-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR