2,2-Dimethyl-3-{[4-(4-nitrophenyl)piperazin-1-yl]carbonyl}cyclopropanecarboxylic acid

Supplier: Matrix Scientific

039037-500MG
101846-698EA 144.9 USD
101846-698
2,2-Dimethyl-3-{[4-(4-nitrophenyl)piperazin-1-yl]carbonyl}cyclopropanecarboxylic acid
2,2-Dimethyl-3-{[4-(4-nitrophenyl)piperazin-1-yl]carbonyl}cyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₁₇H₂₁N₃O₅
MDL Number: MFCD12027542
CAS Number: 1142214-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR