2,2-Dimethyl-3-({4-[(E)-2-phenylvinyl]piperazin-1-yl}carbonyl)cyclopropanecarboxylic acid

Supplier: Matrix Scientific

039036-500MG
101846-696EA 144.9 USD
101846-696
2,2-Dimethyl-3-({4-[(E)-2-phenylvinyl]piperazin-1-yl}carbonyl)cyclopropanecarboxylic acid
2,2-Dimethyl-3-({4-[(E)-2-phenylvinyl]piperazin-1-yl}carbonyl)cyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₁₉H₂₄N₂O₃
MDL Number: MFCD12027541
CAS Number: 1142214-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR