2,2-Dimethyl-3-[(4-pyrimidin-2-ylpiperazin-1-yl)carbonyl]cyclopropanecarboxylic acid

Supplier: Matrix Scientific

039029-500MG
101846-682EA 144.9 USD
101846-682
2,2-Dimethyl-3-[(4-pyrimidin-2-ylpiperazin-1-yl)carbonyl]cyclopropanecarboxylic acid
2,2-Dimethyl-3-[(4-pyrimidin-2-ylpiperazin-1-yl)carbonyl]cyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₁₅H₂₀N₄O₃
MDL Number: MFCD12027534
CAS Number: 1142214-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR