3-[(4-Ethylpiperazin-1-yl)carbonyl]-2,2-dimethylcyclopropanecarboxylic acid

Supplier: Matrix Scientific

039025-500MG
101846-674EA 217.35 USD
101846-674
3-[(4-Ethylpiperazin-1-yl)carbonyl]-2,2-dimethylcyclopropanecarboxylic acid
3-[(4-Ethylpiperazin-1-yl)carbonyl]-2,2-dimethylcyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₂₂N₂O₃
MDL Number: MFCD12027530
CAS Number: 1142214-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR