(2E)-1-(4-Aminophenyl)-3-(2,3-dimethoxyphenyl)prop-2-en-1-one

Supplier: Matrix Scientific

038990-500MG
101846-604EA 144.9 USD
101846-604
(2E)-1-(4-Aminophenyl)-3-(2,3-dimethoxyphenyl)prop-2-en-1-one
(2E)-1-(4-Aminophenyl)-3-(2,3-dimethoxyphenyl)prop-2-en-1-one

Hazard: Irritant

Formula: C₁₇H₁₇NO₃
MDL Number: MFCD09559389
CAS Number: 807642-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR