(2E)-1-(4-Aminophenyl)-3-(2,6-dichlorophenyl)prop-2-en-1-one

Supplier: Matrix Scientific

038989-500MG
101846-602EA 144.9 USD
101846-602
(2E)-1-(4-Aminophenyl)-3-(2,6-dichlorophenyl)prop-2-en-1-one
(2E)-1-(4-Aminophenyl)-3-(2,6-dichlorophenyl)prop-2-en-1-one

Hazard: Irritant

Formula: C₁₅H₁₁Cl₂NO
MDL Number: MFCD09559388
CAS Number: 899017-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR