(2E)-1-(4-Aminophenyl)-3-(1H-indol-4-yl)prop-2-en-1-one

Supplier: Matrix Scientific

038980-500MG
101846-584EA 144.9 USD
101846-584
(2E)-1-(4-Aminophenyl)-3-(1H-indol-4-yl)prop-2-en-1-one
(2E)-1-(4-Aminophenyl)-3-(1H-indol-4-yl)prop-2-en-1-one

Hazard: Irritant

Formula: C₁₇H₁₄N₂O
MDL Number: MFCD12027526
CAS Number: 1142214-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR