2-(1,3-Benzoxazol-2-yl)benzoic acid

Supplier: Matrix Scientific

038961-500MG
101846-548EA 217.35 USD
101846-548
2-(1,3-Benzoxazol-2-yl)benzoic acid
2-(1,3-Benzoxazol-2-yl)benzoic acid

Hazard: Irritant

Formula: C₁₄H₉NO₃
MDL Number: MFCD06254279
CAS Number: 104286-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR