2,1,3-Benzothiadiazole-5-carbaldehyde

Supplier: Matrix Scientific

Synonyms: Benzo[c][1,2,5]thiadiazole-5-carbaldehyde

038959-500MG
101846-544EA 387.69 USD
101846-544
2,1,3-Benzothiadiazole-5-carbaldehyde
2,1,3-Benzothiadiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₇H₄N₂OS
MW: 164.1800 g/mol
MDL Number: MFCD00457949
CAS Number: 71605-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR