4-Amino-2-(butylthio)-N-methylbenzamide

Supplier: Matrix Scientific

038958-500MG
101846-542EA 217.35 USD
101846-542
4-Amino-2-(butylthio)-N-methylbenzamide
4-Amino-2-(butylthio)-N-methylbenzamide

Hazard: Irritant

Formula: C₁₂H₁₈N₂OS
MDL Number: MFCD12027521
CAS Number: 921076-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR