4-Amino-2-(butylthio)benzamide

Supplier: Matrix Scientific

038954-500MG
101846-534EA 217.35 USD
101846-534
4-Amino-2-(butylthio)benzamide
4-Amino-2-(butylthio)benzamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂OS
MDL Number: MFCD12027517
CAS Number: 920483-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR