1-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

038942-500MG
101846-510EA 217.35 USD
101846-510
1-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde ≥95%
1-Methyl-1H-pyrrolo[2,3-b]pyridine-3-carbaldehyde

Hazard: Irritant

Formula: C₉H₈N₂O
MDL Number: MFCD11054209
CAS Number: 171919-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR