N-(1H-Pyrrol-2-ylmethyl)cyclopropanamine

Supplier: Matrix Scientific

038931-500MG
101846-488EA 217.35 USD
101846-488
N-(1H-Pyrrol-2-ylmethyl)cyclopropanamine
N-(1H-Pyrrol-2-ylmethyl)cyclopropanamine

Hazard: Irritant

Formula: C₈H₁₂N₂
Melting Pt: 119…120 °C
MDL Number: MFCD12027502
CAS Number: 643008-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR