[1-(2-Aminophenyl)ethyl](pyridin-3-ylmethyl)amine

Supplier: Matrix Scientific

038930-500MG
101846-486EA 217.35 USD
101846-486
[1-(2-Aminophenyl)ethyl](pyridin-3-ylmethyl)amine
[1-(2-Aminophenyl)ethyl](pyridin-3-ylmethyl)amine

Hazard: Irritant

Formula: C₁₄H₁₇N₃
MDL Number: MFCD12027501
CAS Number: 1142214-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR