2-[(4-Aminophenyl)thio]-N-[5-(ethylthio)-2-thienyl]acetamide

Supplier: Matrix Scientific

038925-500MG
101846-480EA 144.9 USD
101846-480
2-[(4-Aminophenyl)thio]-N-[5-(ethylthio)-2-thienyl]acetamide
2-[(4-Aminophenyl)thio]-N-[5-(ethylthio)-2-thienyl]acetamide

Hazard: Irritant

Formula: C₁₄H₁₆N₂OS₃
MDL Number: MFCD09669578
CAS Number: 931300-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR