N-{4-[(Acetylamino)sulfonyl]phenyl}-2-[(4-aminophenyl)thio]propanamide

Supplier: Matrix Scientific

038922-500MG
101846-474EA 144.9 USD
101846-474
N-{4-[(Acetylamino)sulfonyl]phenyl}-2-[(4-aminophenyl)thio]propanamide
N-{4-[(Acetylamino)sulfonyl]phenyl}-2-[(4-aminophenyl)thio]propanamide

Hazard: Irritant

Formula: C₁₇H₁₉N₃O₄S₂
MDL Number: MFCD09669495
CAS Number: 931293-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR