2-[(4-Aminophenyl)thio]-N-[5-(ethylthio)-1,3,4-thiadiazol-2-yl]propanamide

Supplier: Matrix Scientific

038913-500MG
101846-460EA 144.9 USD
101846-460
2-[(4-Aminophenyl)thio]-N-[5-(ethylthio)-1,3,4-thiadiazol-2-yl]propanamide
2-[(4-Aminophenyl)thio]-N-[5-(ethylthio)-1,3,4-thiadiazol-2-yl]propanamide

Hazard: Irritant

Formula: C₁₃H₁₆N₄OS₃
MDL Number: MFCD09669477
CAS Number: 931292-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR