2-[(4-Aminophenyl)thio]-N-(2,3,5,6-tetrafluorophenyl)propanamide

Supplier: Matrix Scientific

038908-500MG
101846-450EA 144.9 USD
101846-450
2-[(4-Aminophenyl)thio]-N-(2,3,5,6-tetrafluorophenyl)propanamide
2-[(4-Aminophenyl)thio]-N-(2,3,5,6-tetrafluorophenyl)propanamide

Hazard: Irritant

Formula: C₁₅H₁₂F₄N₂OS
MDL Number: MFCD09561572
CAS Number: 913244-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR