2-[(4-Aminophenyl)thio]-N-(4-nitrophenyl)propanamide

Supplier: Matrix Scientific

038907-500MG
101846-448EA 144.9 USD
101846-448
2-[(4-Aminophenyl)thio]-N-(4-nitrophenyl)propanamide
2-[(4-Aminophenyl)thio]-N-(4-nitrophenyl)propanamide

Hazard: Irritant

Formula: C₁₅H₁₅N₃O₃S
MDL Number: MFCD09561159
CAS Number: 899589-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR