2-[(4-Aminophenyl)thio]-N-(2,5-dimethylphenyl)propanamide

Supplier: Matrix Scientific

038900-500MG
101846-434EA 144.9 USD
101846-434
2-[(4-Aminophenyl)thio]-N-(2,5-dimethylphenyl)propanamide
2-[(4-Aminophenyl)thio]-N-(2,5-dimethylphenyl)propanamide

Hazard: Irritant

Formula: C₁₇H₂₀N₂OS
MDL Number: MFCD09560997
CAS Number: 899588-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR