2-[(4-Aminophenyl)thio]-N-(1,3-benzodioxol-5-ylmethyl)propanamide

Supplier: Matrix Scientific

038893-500MG
101846-420EA 144.9 USD
101846-420
2-[(4-Aminophenyl)thio]-N-(1,3-benzodioxol-5-ylmethyl)propanamide
2-[(4-Aminophenyl)thio]-N-(1,3-benzodioxol-5-ylmethyl)propanamide

Hazard: Irritant

Formula: C₁₇H₁₈N₂O₃S
MDL Number: MFCD09561330
CAS Number: 913250-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR