2-[(4-Aminophenyl)thio]-N-(4-methylphenyl)propanamide

Supplier: Matrix Scientific

038890-500MG
101846-414EA 144.9 USD
101846-414
2-[(4-Aminophenyl)thio]-N-(4-methylphenyl)propanamide
2-[(4-Aminophenyl)thio]-N-(4-methylphenyl)propanamide

Hazard: Irritant

Formula: C₁₆H₁₈N₂OS
MDL Number: MFCD09560915
CAS Number: 899016-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR